Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1, Mouse, Clone: 1F1, Abnova™

Catalog No. 89014322 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-322 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-322 Supplier Abnova Corporation Supplier No. H00056913M01
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant C1GALT1.

The common core 1 O-glycan structure Gal-beta-1-3GalNAc-R is a precursor for many extended mucin-type O-glycan structures in animal cell surface and secreted glycoproteins. Core 1 is synthesized by the transfer of Gal from UDP-Gal to GalNAc-alpha-1-R by core 1 beta-3-galactosyltransferase (C1GALT1) (Ju et al., 2002 [PubMed 11677243]).[supplied by OMIM

Sequence: ETFHPFVPEHHLIKGYLPRTFWYWNYNYYPPVEGPGCCSDLAVSFHYVDSTTMYELEYLVYHLRPYGYLYRYQPTLPERILKEISQANKNEDTKVKLGNP

Specifications

Antigen core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant C1GALT1.
Formulation PBS with no preservative; pH 7.4
Gene C1GALT1
Gene Accession No. NM_020156
Gene Alias C1GALT/T-synthase
Gene Symbols C1GALT1
Host Species Mouse
Immunogen C1GALT1 (NP_064541, 264 a.a. ∼ 363 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 56913
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.