Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-C7orf38 Matched Antibody Pair, Abnova

Catalog No. 89017454 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-454 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-454 Supplier Abnova Corporation Supplier No. H00221786AP21
Add to Cart
Add to Cart

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-C7orf38 (100μg); Mouse polyclonal anti-C7orf38 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse.

ELISA

Specifications

Antigen C7orf38
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human C7orf38.
Gene C7orf38
Gene Alias DKFZp727G131/FLJ36794
Gene Symbols C7orf38
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221786
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-C7orf38 (100 μg) 2. Detection antibody: mouse polyclonal anti-C7orf38 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.