Learn More
Advanced ImmunoChemical, Inc. ANTI-CA-125 MAB
Supplier:  Advanced ImmunoChemical, Inc. 2CA1251MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with CA125 antigen purified from human ovarian carcinoma (MW > 1 MDa).Specificity:CA125 antigen.MAbs X52cc, X75, X325: epitope specificity group B (ISOBM classification) similar to M-11.MAb X306cc: epitope specificity group A (ISOBM classification) similar to OC125.MAb Isotypes:IgG1 for all MAbsApplications:Detection of CA125 antigen. Suggested pairs for sandwich immunoassay (capture-detection):X306cc – X52ccX306cc – X325X306cc – X75All MAbs are working in Western blotting.paraffin embedded tissue.Purification:Ion exchange chromatographyPresentation:PBS, pH 7.4, 0.09 % sodium azide (NaN3)0.15 M NaCl, 10 mM Tris, pH 7.5, 0.05% sodium azide (NaN3) for X75, X325Storage Conditions:+ 4 °C (+2°C…+8°C allowed).
Catalog No. NC3565729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.