Learn More
Advanced ImmunoChemical, Inc. ANTI-CA-19-9 MAB
Supplier:  Advanced ImmunoChemical, Inc. 2CA191 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/Monoclonal antibodies / Tumor Markers / Monoclonal mouse anti-CA19-9. MAbs:X91 Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with affinity purified human CA19-9 antigen. Specificity: CA19-9 antigen. MAb Isotypes: IgM for MAb X91Applications: Detection of CA19-9 antigen in immunoassays. MAbs are working in immunohistochemistry. Purification: Ion exchange chromatography. Presentation: 0.15 M NaCl, 10 mM Tris, pH 7.5, 0.05% sodium azide (NaN3). Storage Conditions:+ 4 °C.
Catalog No. NC3566211