Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Channel Cav3.1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5491200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5491200UL 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5491200UL Supplier MilliporeSigma Supplier No. AB5491200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Calcium Channel Cav3.1 Antibody is validated for use in western blotting for the detection of Calcium Channel Cav3.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcium Channel Cav3.1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.05% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).
Gene Symbols CACNA1G; Ca(V)T.1; KIAA1123; Cav3.1; Cav3.1c; NBR13; MGC117234
Host Species Rabbit
Immunogen Highly purified peptide corresponding to residues 1-22 of rat Cav3.1 (Accession O54898).
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_018896.3; NM_198376.1; NM_198377.1; NM_198378.1; NM_198379.1; NM_198380.1; NM_198382.1; NM_198383.1; NM_198384.1; NM_198385.1; NM_198386.1; NM_198387.1; NM_198388.1; NM_198396.1; NM_198397.1
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.