Learn More
Advanced ImmunoChemical, Inc. ANTI-CANINE NT-PROBNP MAB
Supplier:  Advanced ImmunoChemical, Inc. 2CNT1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with: recombinant canine NT-proBNP with tag expressed in E. coli (CaNT90) recombinant canine NT-proBNP expressed in E. coli as a fusion protein with carrier (CaNT89, CaNT19, CaNT46, CaNT925) synthetic peptides corresponding to 3-26 a.a.r., 13-33 a.a.r., and 64- 80 a.a.r. (CaNT49, CaNT53) of canine NT-proBNPSpecificity:Canine NT-proBNP and proBNP.MAb Isotypes:IgG1 for MAbs CaNT89, CaNT90, CaNT19, CaNT46, CaNT49, CaNT53Applications:Quantification of canine NT-proBNP in plasma of dogs. Recommended pairs for sandwich immunoassay (capture-detection):CaNT90 – CaNT89CaNT19 – CaNT89CaNT90 – CaNT53.
Catalog No. NC3566215
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.