Learn More
Advanced ImmunoChemical, Inc. ANTI-CANINE PARVOVIRUS MAB
Supplier:  Advanced ImmunoChemical, Inc. 5PRVC1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:5G7, 8H7, 2A10, 3G3, 3H6Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with canine parvovirus.Specificity:Canine parvovirusMAb Isotypes:IgG2a for MAbs 5G7, 8H7, 2A10, 3G3. IgG3 for MAb 3H6Applications:All MAbs are working in ELISA. MAbs 5G7 and 8H7 are also working in immunodiffusion, haemagglutinin inhibition and lateral flow.Recommended pairs for sandwich immunoassay (capture – detection): 5G7 – 8H7, 3G3 – 8H7, 3H6 – 8H7, 5G7 – 3H6, 3G3 – 3H6, 2A10 – 3H6, 5G7 – 2A10, 3G3 – 2A10Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566230