Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase E Mouse, Unlabeled, Clone: 35, BD

Catalog No. BDB610759 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610759 150 μg
BDB610758 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610759 Supplier BD Biosciences Supplier No. 610759
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carboxypeptidase E (CPE), also known as carboxypeptidase H and enkephalin convertase, is found as both a membrane-bound and a soluble glycoprotein in neuroendocrine tissues and adrenal-gland chromaffin granules. The C-terminus forms an amphiphilic α-helix, suggesting that this region is responsible for the membrane-bound form. Evidence suggests the active form of CPE is located in the secretory vesicles. CPE appears to have several functions. It is an exopeptidase that cleaves neuropeptides with C-terminal basic amino acids, producing an active form of the peptide. It has also been proposed that membrane-bound CPE is a sorting receptor for regulated secretory pathway (RSP) proteins in the TGN pituitary Golgi and secretory granule membranes. RSP proteins primarily consist of hormones and neuropeptides. Mice that carry a mutation in the CPE gene Cpe[fat] display endocrine disorders such as obesity, infertility, and hyperproinsulinemia. Furthermore, the same endocrine disorders are observed in Cpe[fat] mice where the CPE gene has been effaced by antisense RNA.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxypeptidase E
Applications Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Carboxypeptidase E aa. 49-200
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.