Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase Iε Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610445 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610445 50 μg
BDB610446 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610445 Supplier BD Biosciences Supplier No. 610445
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Casein Kinase I? (CKIε) is a product of a family of proteins involved in the regulation of various cytoplasmic and nuclear processes including DNA replication and repair. CKIε is a 47.3kDa serine/threonine kinase that plays a role similar to previously described CKI isoforms. It phosphorylates known CKI substrates, including a CKI-specific peptide. In addition, it is inhibited by CKI-7, an inhibitor of other CKI isoforms. CKIε contains a core kinase domain of 285 amino acids which is 53-98% identical to other isoforms and is most similiar in structure to CKIδ. Since various human and yeast CKI proteins exhibit both sequence and functional similarities, CKIε may be involved in mammalian DNA metabolism.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Casein Kinase Iε
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Casein Kinase Iε aa. 248-414
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.