Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR1, APC, Clone: 53504, R&D Systems™

Catalog No. FAB145A025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB145A025 25 Tests
FAB145A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB145A025 Supplier R&D Systems Supplier No. FAB145A025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Host Species: Mouse
Clone: 53504
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: Swiss 3T3 mouse cell line transfected with human CCR1.

Applications:

Flow Cytometry

Specifications

Antigen CCR1
Applications Flow Cytometry
Classification Monoclonal
Clone 53504
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P32246
Gene Alias C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Gene Symbols CCR1
Host Species Mouse
Immunogen Swiss 3T3 mouse cell line transfected with human CCR1, Accession # P32246
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1230
Test Specificity Detects human CCR1. Reacts specifically with human CCR1 transfectants and not the parental cell line in flow cytometry. No cross-reactivity with CCR5 transfectants is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.