Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CCR10 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4712396
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3478A025 25 Tests
FAB3478A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB3478A025 Supplier R&D Systems Supplier No. FAB3478A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR10/GPR2
Applications Flow Cytometry
Classification Monoclonal
Clone 314305
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P46092
Gene Alias CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2
Gene Symbols CCR10
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with human CCR10 Gln8-Asn362 Accession # P46092
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 2826
Test Specificity Detect human CCR10. Stains human CCR10-transfected cells but not irrelevant transfectants.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.