Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CCR6, Alexa Fluor 700, Clone: 140706, R&D Systems™

Catalog No. FAB590N025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB590N025 25 Tests
FAB590N100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB590N025 Supplier R&D Systems Supplier No. FAB590N025
This item is not returnable. View return policy

Host Species: Rat
Clone: 140706
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6. Met1-Met367

Applications:

Flow Cytometry

Specifications

Antigen CCR6
Applications Flow Cytometry
Classification Monoclonal
Clone 140706
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. O54689
Gene Alias BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22
Gene Symbols CCR6
Host Species Rat
Immunogen RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6, Met1-Met367, Accession # O54689
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1235
Test Specificity Detects mouse CCR6-transfected rat RBL cells but not the parent cell line or cells transfected with mouse CCR5.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.