Learn More
anti-CCR6, Alexa Fluor 700, Clone: 140706, R&D Systems™
Description
Host Species: Rat
Clone: 140706
Isotype: IgG2A
Species Reactivity: Ms
Immunogen: RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6. Met1-Met367
Applications:
Flow Cytometry
Specifications
Specifications
| Antigen | CCR6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 140706 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Gene Accession No. | O54689 |
| Gene Alias | BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 |
| Gene Symbols | CCR6 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.