Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat CCR8 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4712467
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1429A025 25 Tests
FAB1429A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1429A025 Supplier R&D Systems Supplier No. FAB1429A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

CCR8 Monoclonal specifically detects CCR8 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR8
Applications Flow Cytometry
Classification Monoclonal
Clone 191704
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P51685
Gene Alias CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR8, CCR-8, CD198, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, ChemR1, CKRL1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCL1, GPRCY6, GPR-CY6, TER-1, TER1CC chemokine receptor CHEMR1
Gene Symbols CCR8
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with human CCR8 Met1-Leu355 Accession # P51685
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1237
Test Specificity Detects human and rat CCR8. Neutralizes the bioactivity of human CCR8 and specifically stains human CCR8-transfected cell lines but not the parent cell lines.
Target Species Human, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.