Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD109 Mouse anti-Human, Unlabeled, Clone: TEA 2/16, BD

Catalog No. BDB556039 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556039 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556039 Supplier BD Biosciences Supplier No. 556039
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 150-170 kDa glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed on activated T cells, myeloid progenitor (CD34+) and mature myeloid lineage cells (monocytes, granulocytes, platelets), but not on CD34+ lymphoid progenitor cells. CD109 is also expressed on vein and artery endothelial cells. The expression of CD109 is upregulated on PHA-stimulated T cells. CD109 has been reported to be present on long-term adult bone marrow cultured cells, where it is co-expressed with CD34 and CD90. Its biological role in hematopoiesis has not been fully elucidated.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD109
Applications Flow Cytometry
Classification Monoclonal
Clone TEA 2/16
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.