Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD110 Mouse anti-Human, Unlabeled, Clone: 1.78.1, BD

Catalog No. BDB562137 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562137 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562137 Supplier BD Biosciences Supplier No. 562137
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1.78.1 monoclonal antibody specifically binds to the human Thrombopoietin Receptor (TPO-R) that is also know as the Myeloproliferative leukemia protein (c-Mpl) or CD110. CD110 is a type I transmembrane protein and a member of the hematopoietin receptor family. It is expressed on hematopoietic stem cells, a subfraction of hematopoietic precursor cells, cells of the megakaryocytic lineage and platelets. CD110 serves as a receptor for thrombopoietin. Upon binding of thrombopoietin to CD110, megakaryocyte proliferation and differentiation is induced and stem cells are protected from apoptosis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD110
Applications Western Blot
Classification Monoclonal
Clone 1.78.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description C-MPL; MPL; MPLV; TPOR; Thrombopoietin receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human c-Mpl extracellular domain
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.