Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD116 Mouse anti-Human, PE, Clone: HGMCSFR-M1, BD

Catalog No. BDB551373 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551373 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551373 Supplier BD Biosciences Supplier No. 551373
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The hGMCSFR-M1 antibody reacts with the subunit (GM-CSFR) of the human Granulocyte-Macrophage Colony-Stimulating Factor Receptor complex. This 75-85 kD subunit is also known as CD116. The hGMCSFR-M1 antibody was first clustered at the Fifth International Workshop on Human Leucocyte Differentiation Antigens. The GM-CSFR subunit associates with the 120-140 kD βc subunit (common subunit, CD131), that is shared with the receptors for interleukins IL-3 and IL-5. Both of the chains of the GM-CSFR complex are involved in ligand binding and intracellular signaling. The α chain appears to transmit most of the biological signals. CD116 is expressed by a variety of myeloid cell lines, hematopoietic and non-hematopoetic tumor cells, and normal cell types including monocytes, macrophages, neutrophils, eosinophils, myeloid dendritic cells, endothelial cells, fibroblasts, and placental trophoblasts. Lymphocytes are negative for GM-CSFR expression. Reports suggest that GM-CSFR plays a role in myeloid lineage growth and differentiation. The immunogen used to generate the hGMCSFR-M1 hybridoma was recombinant human GM-CSFR.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD116
Applications Flow Cytometry
Classification Monoclonal
Clone HGMCSFR-M1
Concentration 0.2mg/mL
Conjugate PE
Description CSF2RA; GM-CSF Receptor alpha; GM-CSFRα; GMCSFRA; GMR, SMDP4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant human GM-CSFR
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.