Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11b Mouse anti-Rat, APC, Clone: WT.5, BD

Catalog No. BDB562102 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562102 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562102 Supplier BD Biosciences Supplier No. 562102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The WT.5 monoclonal antibody specifically binds to CD11b, the α subunit of Mac-1 found on neutrophils and myeloid cells, but not on lymphocytes. Mac-1 expression is rapidly upregulated on neutrophils after activation, in the same time period that CD62L (L-selectin) is shed from the cell surface. Mac-1 mediates adhesion to ICAM-1 (CD54) and C3bi. Neutrophil binding to ICAM-1 is blocked by mAb WT.5.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone WT.5
Concentration 0.2mg/mL
Conjugate APC
Description Itgam; Integrin αM chain; Mac-1 α chain; Integrin beta 2 alpha subunit
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rt neutrophils
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgA κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.