Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD124 Mouse anti-Human, PE, Clone: HIL4R-M57, BD

Catalog No. BDB552178 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552178 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552178 Supplier BD Biosciences Supplier No. 552178
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The hIL4R-M57 antibody specifically binds to the α subunit (IL-4R α ) of the human Interleukin-4 Receptor complex which is also known as CD124. The human IL-4R α , also known as B cell stimulatory factor 1 receptor (BSF-1 receptor), is a 140 kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and non-hematopoietic cells and cell lines. The cell surface IL-4R α chain binds IL-4 with high affinity and associates with either the common γchain (IL-4R α / γα exists in soluble form in biological fluids. In contrast to mice, in humans no distinct mRNA coding for sIL-4R has been described, suggested that human sIL4-R is exclusively produced by proteolytic cleavage of the cell surface receptor. The serum levels of soluble IL-4R α appear to elevate in pathological situations such as allergy and parasitic infections. Depending on the ratios of IL-4 and sIL-4R α present in the local milieu, the sIL-4R α may augment or antagonize the activities of IL-4. The immunogen used to generate the hIL4R-M57 hybridoma was soluble human IL-4R.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD124
Applications Flow Cytometry
Classification Monoclonal
Clone HIL4R-M57
Conjugate PE
Description IL-4 Receptor α Chain, CD124
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Soluble Human IL-4 Receptor
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.