Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD130 Mouse anti-Human, Alexa Fluor 647, Clone: AM64, BD

Catalog No. BDB564151 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564151 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564151 Supplier BD Biosciences Supplier No. 564151
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AM64 monoclonal antibody specifically binds to CD130 which is also known as gp130 (Membrane glycoprotein 130), IL6ST (Interleukin-6 signal transducer), and IL6RB/ IL-6Rβ (Interleukin-6 Receptor beta). CD130 is a 130 kDa type 1 membrane glycoprotein that belongs to the hemopoietic cytokine receptor superfamily. CD130 serves as a common signaling receptor for several cytokine receptor complexes including those that mediate the biological effects of IL-6, IL-11, IL-27, Cardiotrophin-1 (CT-1), Ciliary neurotrophic factor (CNTF), Leukemia inhibitory factor (LIF), and Oncostatin M (OSM). CD130 is widely expressed on a variety of cell types including T cells, activated B cells, monocytes, plasma cells, and endothelia. The AM64 antibody can be used in flow cytometry, immunoprecipitation, western blot, and immunohistochemistry on frozen sections.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD130
Applications Flow Cytometry
Classification Monoclonal
Clone AM64
Conjugate Alexa Fluor 647
Description IL6ST; IL6RB; IL-6Rβ; Interleukin-6 Receptor beta; gp130
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Human CD130
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.