Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD141 Mouse anti-Human, BB515, Clone: 1A4, BD

Catalog No. BDB565084 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565084 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565084 Supplier BD Biosciences Supplier No. 565084
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1A4 monoclonal antibody specifically binds to a 75 kDa, a single chain, type I membrane glycoprotein referred to as thrombomodulin and fetomodulin. CD141 is expressed on endothelial cells, smooth muscle cells, epithelial cells, synovial lining cells, and keratinocytes. It is also found on megakaryocytes, dendritic cells, peripheral blood moncytes, and neutrophils, but not on lymphocytes. Reports indicate that CD141 plays an important role in Protein C activation and the initiation of the Protein C anticoagulant pathway. Thrombin loses its procagulant function when it binds to CD141, and the CD141/thrombin complex is able to activate Protein C.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD141
Applications Flow Cytometry
Classification Monoclonal
Clone 1A4
Conjugate Brilliant Blue 515 (BB515)
Description Thrombomodulin; TM; THBD; THRM; AHUS6; Fetomodulin; BDCA-3
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Purified Human Thrombomodulin
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.