Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD153 Mouse anti-Human, Unlabeled, Clone: D2-1173, BD

Catalog No. BDB550052 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550052 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550052 Supplier BD Biosciences Supplier No. 550052
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD30 ligand (CD153) a 40 kDa type II membrane glycoprotein expressed mainly on activated T cells, as well as on activated macrophages, neutrophils and both normal and malignant B cells. CD30 ligand is a member of the tumor necrosis factor (TNF) superfamily. In addition to TNF, this family also includes lymphotoxin α (LT α ), LT β , CD154, CD95L, CD70, CD134 ligand, CDw137 ligand, and TRAIL. Results of in vitro studies have shown that CD30 mediates a variety of activation and differentiation, or apoptotic signals. Recently, it has been reported that CD30-deficient mice have elevated numbers of thymocytes and show a gross defect in negative but not positive selection during T cell differentiation, suggesting an important role for CD30/CD30L interactions in the deletion of autoreactive T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD153
Applications Flow Cytometry
Classification Monoclonal
Clone D2-1173
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CD30L, CD30 ligand
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.