Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158a Mouse anti-Human, Brilliant Violet 421, Clone: HP-3E4, BD

Catalog No. BDB564318 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564318 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564318 Supplier BD Biosciences Supplier No. 564318
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HP-3E4 monoclonal antibody recognizes CD158a. CD158a is a 58 kDa membrane glycoprotein, member of the killer-inhibitory receptor (KIR). The KIR family consists of transmembrane glycoproteins of the Ig superfamily expressed on <1-8% of NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. CD158a is an antigen which recognizes HLA-C molecules (-Cw*0401 and -Cw*1503). This antibody is suitable for staining acetone-fixed, frozen tissue sections. It was reported that this clone also crossreacted with NKAT8 (CD158i) transfected cells.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158a
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3E4
Conjugate Brilliant Violet 421
Description KIR2DL1; KI2L1; NKAT; NKAT-1; NKAT1; KIR-K64; KIR221; p58.1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.