Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15s Mouse anti-Human, Unlabeled, Clone: CSLEX1, BD

Catalog No. BDB551344 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551344 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551344 Supplier BD Biosciences Supplier No. 551344
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CSLEX1 monoclonal antibody specifically binds to CD15s. This anti-Sialyl-Le[x] monoclonal antibody is specific for the α 2-3 sialosylated form of lacto-N-fucopentaose III, sialyl Lex (Sle[x]). Sle[x] is expressed on granulocytes, monocytes and both normal and tumor cells of diverse origin. It has been shown to be a ligand for both endothelial leukocyte adhesion molecule-1 (ELAM-1 or E-selectin), and granule membrane protein-140 (GMP-140 or P-selectin).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15s
Applications Flow Cytometry
Classification Monoclonal
Clone CSLEX1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Sialyl Lewis X
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Membrane proteins from human tomach adenocarcinoma tissue
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.