Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16 Mouse anti-Human, APC, Clone: B73.1, BD

Catalog No. BDB561304 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561304 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561304 Supplier BD Biosciences Supplier No. 561304
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The B73.1 monoclonal antibody specifically binds to CD16, the 50-70 kDa low affinity receptor for IgG, IgG Fc receptor III (Fc γRIII) . CD16 is expressed on NK cells, neutrophils, and on a subset of T cells from certain individuals. The B73.1 antibody binds to CD16-positive neutrophils with lower intensity when compared with some other CD16-specific antibodies. A variable number of CD16-positive lymphocytes coexpress either the CD57 antigen or low-density CD8 antigen or both. The B73.1 antibody can block Fc receptor functions mediated by CD16.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16
Applications Flow Cytometry
Classification Monoclonal
Clone B73.1
Conjugate APC
Description IgG Fc receptor III; IGFR3; FCG3; FCGR3; FCGRIII; FcγRIII
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.