Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Rat anti-Mouse, Unlabeled, Clone: 2.4G2, BD

Catalog No. BDB553141 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553141 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553141 Supplier BD Biosciences Supplier No. 553141
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse Fc γ III and Fc γ II receptors. It has also been reported to bind the Fc γ I receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the Fc γ III and Fc γ II, and possibly Fc γ I, receptors in vitro and in vivo . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16/CD32
Applications Blocking Assay, Flow Cytometry
Classification Monoclonal
Clone 2.4G2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description FcγRIII/FcγRII; Fcgr3/Fcgr2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse BALB/c Macrophage J774 Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.