Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD178 Mouse anti-Human, Unlabeled, Clone: G247-4, BD

Catalog No. BDB556387 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556387 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556387 Supplier BD Biosciences Supplier No. 556387
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fas (APO-1, CD95) is a 45 kD cell surface protein that mediates apoptosis when crosslinked with agonistic anti-Fas antibodies or Fas ligand (FasL). Fas belongs to the TNF (tumor necrosis factor)/NGF (nerve growth factor) receptor family, and is expressed in various tissue and cells including the thymus, liver, ovary and lung. FasL is a member of the TNF cytokine family that induces apoptosis by binding to Fas, its cell-surface receptor. FasL may exist in both membrane and soluble forms and expressed on activated T cells, NK cells, and other “immunologically privileged” sites. Both Fas and FasL are thought to play an important role in the apoptotic processes that take place during T cell development.

G247-4 recognizes human FasL. It recognizes both the membrane bound (FasL) and soluble (sFasL) forms. A recombinant protein containing the external domain of human FasL was used as immunogen. FasL and sFasL migrate at reduced molecular weights of 40 and 26 kD, respectively. However, the molecular weights observed in a particular sample may vary according to FasL and sFasL glycosylation and breakdown patterns as described in Tanaka et al. For example, FasL may migrate as a doublet of 40 and 42 kD.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD178
Applications Western Blot
Classification Monoclonal
Clone G247-4
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Fas Ligand, CD95 Ligand
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human FasL
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.