Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD249 Mouse anti-Human, PE, Clone: 2D3/APA, BD

Catalog No. BDB564533 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564533 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564533 Supplier BD Biosciences Supplier No. 564533
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2D3 monoclonal antibody specifically binds to CD249. CD249 is encoded by ENPEP and is also known as Aminopeptidase A (APA/AP-A), Glutamyl aminopeptidase (EAP), and gp160. CD249 is expressed by epithelial and endothelial cells in a variety of tissues including those that comprise the proximal tubules and glomeruli of the kidneys. It is also expressed by early B-lineage cells and cell lines. CD249 belongs to the peptidase M1 family and catalyzes the release of N-terminal glutamate (and to a lesser degree, aspartate) from peptides. This ectoenzyme catalyzes the conversion of angiotensin II to angiotensin III which is important in the local regulation of blood pressure. It may also play roles in the regulation of angiogenesis and development of B cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD249
Applications Flow Cytometry
Classification Monoclonal
Clone 2D3/APA
Conjugate PE
Description EAP, Aminopeptidase A, Glutamyl aminopeptidase, APA, AP-A, gp160, ENPEP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Aminopeptidase A Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.