Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25 Mouse anti-Human, Unlabeled, Clone: M-A251, BD

Catalog No. BDB555430 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555430 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555430 Supplier BD Biosciences Supplier No. 555430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD25, a 55kDa glycoprotein known as the low-affinity interleukin-2 receptor (IL-2R). CD25 is expressed on activated lymphocytes (T and B) and monocytes, and associates with the p75 high-affinity receptor chain to form the IL-2R complex. The CD25 molecule reveals three epitope regions: A, B, and C. M-A251 antibody recognizes epitope region B. CD25 expression on lymphocytes is upregulated by anti-CD3 or PHA stimulation. Soluble IL-2R is produced following inflammatory responses. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25
Applications Flow Cytometry
Classification Monoclonal
Clone M-A251
Concentration 0.5mg/mL
Conjugate Unconjugated
Description IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.