Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274 Mouse anti-Human, APC, Clone: MIH1, BD

Catalog No. BDB563741 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563741 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563741 Supplier BD Biosciences Supplier No. 563741
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH1 monoclonal antibody specifically binds to CD274, also known as Programmed Death Ligand 1 (PD-L1) and B7 Homolog 1 (B7-H1). CD274/PD-L1 and CD273/PD-L2 are members of the B7 family within the Immunoglobulin superfamily. These type I transmembrane glycoproteins function as ligands for CD279, likewise known as the Programmed Death 1 (PD-1) receptor. CD274 is expressed on activated dendritic cells, macrophages and T cells. Studies show some overlapping functions of CD274 and CD273 and indicate an important role for CD279 and its ligands in regulating T and B cell responses. CD274 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, keratinocytes, and carcinomas. The MIH1 antibody is reportedly a blocking antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274
Applications Flow Cytometry
Classification Monoclonal
Clone MIH1
Conjugate APC
Description B7-H1; Programmed death ligand 1; PD-L1; PDCD1 ligand 1; PDCD1L1; PDCD1LG1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human B7-H1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.