Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Hamster anti-Mouse, PE, Clone: MCD30.1, BD

Catalog No. BDB553825 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553825 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553825 Supplier BD Biosciences Supplier No. 553825
Only null left
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The mCD30.1 antibody reacts with CD30, a member of the Tumor Necrosis Factor Receptor (TNFR) family. The CD30 molecule is predominantly expressed by activated T lymphocytes, with its expression peaking at day 4-5 on spleen cells activated with plate-bound anti-CD3e antibody. The mCD30.1 antibody reacts with a majority of CD8+ T cells, as well as some CD4+ T cells in these cultures. Expression of CD30 on activated T lymphocytes is regulated by CD28 and cytokines. Its TNF-superfamily ligand, CD30L or CD153, is also expressed on activated T lymphocytes. By northern blot analysis, mouse Cd30 mRNA is detected in the thymus and in 72-hour pokeweed mitogen- and Con A-activated spleen cells, but not in the lung, brain, kidney, liver, bone marrow, unactivated spleen, or 72-hour LPS-activated splenocytes.1 It has also been reported that CD30 is expressed on naive B lymphocytes, it is not detectable after activation, and it starts to return after 72 hours following activation. Reports suggest that signaling through the CD30 molecule may be important in cytokine production by CD8+ CTL lines and may play a role in the regulation of Th1 and Th2 cytokine secretion by CD4+ and CD8+ T cells. It has also been proposed that CD30 plays an important role in the negative selection of thymocytes and protects against autoimmunity. Members of the TNFR family and their ligands are involved in the induction of diverse biological responses in lymphocytes, including differentiation, proliferation, and cellular death. In humans, CD30 was initially identified in Hodgkin and Reed-Sternberg cells in Hodgkin’s disease patients and subsequently was found on neoplastic cells of certain types of non-Hodgkin’s lymphomas.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30
Applications Flow Cytometry
Classification Monoclonal
Clone MCD30.1
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse CD30-IgG1 fusion protein
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.