Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD309 (VEGFR-2) Mouse anti-Human, PE, Clone: 89106, BD

Catalog No. BDB560494 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560494 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560494 Supplier BD Biosciences Supplier No. 560494
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 89106 monoclonal antibody reacts with CD309 (vascular endothelial growth factor r eceptor-2 (VEGFR-2)), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. VEGFR-2, al so known as fetal liver kinase 1 (Flk-1) or kinase insert domain receptor (KDR), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic tissues. In vivo in vitro studies indicate that VEGFR-2 is required for the embryonic development of vascular endothelial and hematopoietic cells. Human cardiac progenitor cells derived from human embryonic stem cells arise from a population of cells that express VEGFR-2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD309
Applications Flow Cytometry
Classification Monoclonal
Clone 89106
Conjugate PE
Description FLK1; Fetal liver kinase 1; KDR; VEGFR2; VEGF Receptor 2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human VEGFR-2
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.