Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human, Alexa Fluor 647, Clone: M89D3, BD

Catalog No. BDB558094 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558094 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558094 Supplier BD Biosciences Supplier No. 558094
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reacts with CD31, also known as platelet endothelial cell adhesion molecule - 1 (PECAM-1), a single chain, type I transmembrane protein of approximately 130 - 140 kDa. CD31 is a member of the immunoglobulin supergene family, its extracellular region is composed of 574 amino acids and contains six C2-type Ig domains, its intracellular region is composed of 118 residues. CD31 (PECAM-1) is expressed on endothelial cells, platelets, monocytes, neutrophils and NK cells. It has also been observed on subsets of T cells, but not on circulating B cells. It has been implicated in a number of cellular phenomena, including vascular wound healing, angiogenesis, and transendothelial migration of leukocytes and platelet aggregation in inflammatory responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone M89D3
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description PECAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.