Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD31/PECAM-1 Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
$138.04 - $288.84
Specifications
Antigen | CD31/PECAM-1 |
---|---|
Clone | 9G11 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB3567F025
|
R&D Systems
FAB3567F025 |
25 Tests |
Each for $138.04
|
|
|||||
FAB3567F100
|
R&D Systems
FAB3567F100 |
100 Tests |
Each for $288.84
|
|
|||||
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human samples. It is validated for Flow Cytometry.Specifications
CD31/PECAM-1 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
5175 | |
Activated HUVEC human umbilical vein endothelial cells | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
9G11 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
Human | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
PECAM1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) E-Selectin, rhICAM-1, -2, -3, rhVCAM-1, or recombinant mouse VCAM-1 is observed. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title