Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD32 Mouse anti-Human, Unlabeled, Clone: FLI8.26, BD

Catalog No. BDB555447 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555447 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555447 Supplier BD Biosciences Supplier No. 555447
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the 40kDa type I membrane glycoprotein, FcγRII, the low affinity receptor for aggregated IgG expressed on B cells, monocytes, granulocytes, and platelets. CD32 mediates endocytosis, cytotoxicity, platelet activation and immunomodulation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: FLI8.26 (2003)
Isotype: IgG2b, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Applications Flow Cytometry
Classification Monoclonal
Clone FLI8.26
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen K562 or FcγRII+ L cell lines
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.