Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 ligand, Mouse, Clone: 3E4, Abnova™

Catalog No. 89000564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-564 Supplier Abnova Corporation Supplier No. H00000959M04
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CD40LG.

The protein encoded by this gene is expressed on the surface of T cells. It regulates B cell function by engaging CD40 on the B cell surface. A defect in this gene results in an inability to undergo immunoglobulin class switch and is associated with hyper-IgM syndrome. [provided by RefSeq

Sequence: MQKGDQNPQIAAHVISEASSKTTSVLQWAEKGYYTMSNNLVTLENGKQLTVKRQGLYYIYAQVTFCSNREASSQAPFIASLCLKSPGRFERILLRAANTHSSAKPCGQQSIHLGGVFELQPGASVFVNVTDPSQVSHGTGFTSFGLLKL

Specifications

Antigen CD40 Ligand
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CD40LG.
Formulation PBS with no preservative; pH 7.4
Gene CD40LG
Gene Accession No. N/A
Gene Alias CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L
Gene Symbols CD40LG
Host Species Mouse
Immunogen CD40LG (NP_000065, 113 a.a.-261 a.a.) partial recombinant protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 959
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.