Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42d Mouse anti-Rat, FITC, Clone: RPM.4, BD

Catalog No. BDB551805 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551805 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551805 Supplier BD Biosciences Supplier No. 551805
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RPM.4 antibody reacts with CD42d (Platelet glycoprotein V, GPV), an 88-kDa glycoprotein which noncovalently associates with GPIb and GPIX to form a receptor for von Willebrand factor on megakaryocytes and resting platelets. The binding sites for von Willebrand factor and thrombin have been localized to the GPIbα chain of the GPI-b-V-IX complex. GPV is cleaved by thrombin to release a 70 kDa soluble fragment, which contains the epitope recognized by mAb RPM.4. Although the rat CD42d protein's structure is similar to that of human CD42d, the RPM.4 antibody does not cross-react with human platelets.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42d
Applications Flow Cytometry
Classification Monoclonal
Clone RPM.4
Concentration 0.5mg/mL
Conjugate FITC
Description Platelet Glycoprotein V
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Wistar rat platelets
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.