Learn More
Abcam Anti-CD44 Antibody, Rabbit Multiclonal, RM1084, 1 mg
Description
- Host species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Purification technique: Affinity purification Protein A
- Applications: WB, ICC/IF, Flow Cyt, IHC-P
- Species reactivity: Human, Mouse, Rat
- Form: Liquid
- Storage buffer: PBS (pH 7.2 to 7.4)
- Other: Carrier free, conjugation ready, BSA and azide free
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Membrane Compatibility | Nitrocellulose, PVDF |
| Form | Liquid |
| Detection Method | Chromogenic, Chemiluminescence, Fluorescence |
| Detection Location | In-Blot Detection, In-Cell Detection |
| Buffer | PBS, pH7.2 to 7.4 |
| Label or Dye | Unconjugated |
| Target Molecule | CD44 |
| Cross Reactivity | Human, Mouse, Rat |
| Formulation | PBS, pH 7.2 to 7.4 |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.