Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 (Pgp-1, H-CAM, Ly24) Mouse anti-Human, PerCP-Cy5.5, Clone: G44-26, BD

Catalog No. BDB560531 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560531 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560531 Supplier BD Biosciences Supplier No. 560531
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G44-26 monoclonal antibody specifically binds to the 80-95 kDa, glycosylated type I transmembrane protein, also known as phagocytic glycoprotein-1 (Pgp-1). CD44 is the receptor for hyaluronic acid. CD44 is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is also called extracellular matrix receptor type III and has functional roles in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using citrate buffer pretreatment. This antibody recognizes the epitope 1 of CD44 antigen according to the HLDA workshop studies.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44 (Pgp-1, H-CAM, Ly24)
Applications Flow Cytometry
Classification Monoclonal
Clone G44-26
Conjugate PerCP-Cyanine5.5
Description Phagocytic glycoprotein 1; Pgp-1; H-CAM; Hermes; ECMR III; HUTCH-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Dibutyryl cAMP-activated 5C2 cells, which were derived by transfecting truncated H2-Aak and H2-Abk cDNA into the Ia-negative BALB/c mouse lymphoma M12.C3
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.