Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Rat, APC-Cy7, Clone: OX-1, BD

Catalog No. BDB561586 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561586 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561586 Supplier BD Biosciences Supplier No. 561586
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-1 antibody recognizes all molecular forms of CD45 (Leukocyte Common Antigen) on all hematopoietic cells except erythrocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family; its intracellular (COOH-terminal) region contains two PTP catalytic domains while the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), and differing levels of glycosylation. The CD45 isoforms detected in rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone OX-1
Concentration 0.2mg/mL
Conjugate APC-Cyanine7
Description Ptprc; Lca; Leucocyte common antigen; RT7; T200
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Leukocyte Common Antigen-enriched Glycoprotein Fraction from Wistar Rat Thymocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.