Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-CD47 Antibody RM1014, Rabbit Multiclonal, BSA and Azide Free, 1 mg

Catalog No. 502655081
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-5081 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-5081 Supplier Abcam Supplier No. AB2841381MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Rabbit multiclonal antibody for CD47 staining in flow cytometry, ICC/IF, IHC, and western blot applications.

  • Applications: WB, ICC/IF, Flow Cyt, IHC-P
  • Host Species: Rabbit
  • Clonality: Multiclonal
  • Isotype: IgG
  • Reactivity: Human
  • Purification: Affinity purification Protein A
  • Form: Liquid
  • Storage Buffer: PBS (pH 7.2 - 7.4)
  • Additional Features: BSA and Azide free; tested in knockout cell lines; tested for Leica Bond™

The product is subject to the following: Abcam Restricted Use Statement

Specifications

For Use With (Equipment) Leica Bond System
Target Molecule CD47
Label or Dye Unconjugated
Form Liquid
Detection Method Chromogenic, Chemiluminescence, Fluorescence
Buffer PBS, pH7.2 to 7.4
Detection Location In-Blot Detection, In-Cell Detection
Cross Reactivity Human
Formulation PBS, pH 7.2 to 7.4
Quantity 1 mg
For Use With (Application) Flow Cytometry, ICC/IF, IHC-P, Western Blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.