Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Rat anti-Mouse, Unlabeled, Clone: MIAP301, BD

Catalog No. BDB555297 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555297 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555297 Supplier BD Biosciences Supplier No. 555297
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The miap301 monoclonal antibody specifically binds to the extracellular domain of CD47, also known as Integrin-Associated Protein (IAP). CD47 is a 50-kDa plasma membrane protein with an extracellular immunoglobulin variable region-like domain, a multiple membrane-spanning segment, and a short C-terminal cytoplasmic tail. IAP is often physically associated with β3 integrins, and it may act as a transducer element in activation mediated by these integrins. CD47 is expressed by a wide variety of cell types, including some cells lacking integrins, such as erythrocytes. The cytoplasmic tail region is expressed as alternatively spliced forms, which are differentially expressed in diverse tissues, suggesting that IAP may have distinct functions in various tissues. CD47 and its ligand, CD172a, are proposed to mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD47
Applications Flow Cytometry
Classification Monoclonal
Clone MIAP301
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Cd47; IAP; Integrin-associated protein; Itgp
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Not reported
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.