Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d (Integrin α4 chain) Mouse anti-Human, Unlabeled, Clone: L25, BD

Catalog No. BDB340976 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340976 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340976 Supplier BD Biosciences Supplier No. 340976
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD49d (Anti–VLA-α4), clone L25, is derived from the fusion of mouse Sp2/0 cells with popliteal lymph node cells from a BALB/c mouse immunized with a CD8+ T-cell line.

CD49d (Anti–VLA-α4) recognizes the 150-kilodalton (kd) α chain of very-late antigen (VLA)-4, a member of the integrin family of cell adhesion molecules. VLA-4, like other integrins, is a noncovalently associated heterodimeric glycoprotein composed of α and β subunits and is involved in cell–cell and cell–extracellular matrix interactions. The β chain of the VLA-4 complex is the CD29 antigen, a 130-kd glycoprotein. The CD29 antigen, also known as the β1 subunit, is common to the VLA family of integrins. When acting as a matrix receptor, the CD49d antigen binds to CS-1, an alternatively spliced domain of fibronectin. When functioning as a cell receptor, the CD49d antigen binds to the vascular cell-adhesion molecule-1 (VCAM-1). The interaction between the CD49d antigen and VCAM-1 is known to play an important role in stabilizing the adhesion of lymphocytes to endothelial cells and in mediating B-lymphocyte precursor/bone marrow stromal cell adhesion. The CD49d antigen, when associated with the β7 integrin, forms a lymphocyte homing receptor for Peyer's patch, binding to the mucosal vascular addressin MAdCAM-1. The CD49d antigen is also involved in CD3-dependent CD4+T-lymphocyte activation via its interaction with fibronectin.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone L25
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen CD8
Purification Method Affinity Purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.