Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 Mouse anti-Human, Unlabeled, Clone: HNK-1, BD

Catalog No. BDB347390 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347390 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347390 Supplier BD Biosciences Supplier No. 347390
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD57, clone HNK-1, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with lymph node cells from BALB/c mice immunized with membrane extracts of the HSB-2 T–lymphoblastoid cell line.

CD57 (Leu-7) recognizes an Mr 110-kdalton (kDa) glycoprotein that is expressed on a subset of natural killer (NK) lymphocytes and T lymphocytes. The antigen is a carbohydrate structure associated with myelin-associated glycoprotein (MAG).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Applications Flow Cytometry
Classification Monoclonal
Clone HNK-1
Concentration 12.5μg/mL
Conjugate Unconjugated
Description Leu-7
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Membrane extracts of the HSB-2 T-lymphoblastoid cell line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.