Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD70 Mouse anti-Human, Unlabeled, Clone: KI-24, BD

Catalog No. BDB555833 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555833 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555833 Supplier BD Biosciences Supplier No. 555833
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The Ki-24 monoclonal antibody specifically binds to human CD70. CD70 is a type II transmembrane glycoprotein and member of the TNF Superfamily. CD70 is also known as Tumor necrosis factor ligand superfamily member 7 (TNFSF7), CD27 ligand (CD27-L, CD27L, CD27LG), and KI-24 antigen. The CD70 antigen immunoprecipitates as five bands (50, 70, 90, 100 and 160 kDa) under non-reducing conditions. CD70 is strongly expressed on Reed-Sternberg cells, some activated T or B cells and Epstein Barr Virus (EBV)-positive lymphoblastoid cell lines. CD70 plays roles in the activation, proliferation and differentiation of B cells and T cells including the enhanced production of cytotoxic T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD70
Applications Flow Cytometry
Classification Monoclonal
Clone KI-24
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG3 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.