Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD71 (Transferrin Receptor) Mouse anti-Human, FITC, Clone: L01.1, BD

Catalog No. BDB347513 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347513 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347513 Supplier BD Biosciences Supplier No. 347513
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD71 antibody, clone L01.1, is derived from the hybridization of P3-X63- Ag8.653 mouse myeloma cells with spleen cells from BALB/c mice immunized with blast cells from pokeweed mitogen–stimulated peripheral blood cells.

The CD71 antibody is specific for the human transferrin receptor, with a molecular weight of 190 kilodaltons (kDa), 90-kDa reduced.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD71 (Transferrin Receptor)
Applications Flow Cytometry
Classification Monoclonal
Clone L01.1
Concentration 100μg/mL
Conjugate FITC
Description Transferrin Receptor
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Blast cells from pokeweed mitogen-stimulated peripheral blood cells
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.