Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD73 Mouse anti-Human, Unlabeled, Clone: AD2, BD

Catalog No. BDB550256 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550256 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550256 Supplier BD Biosciences Supplier No. 550256
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AD2 monoclonal antibody specifically binds to ecto-5’-nucleotidase, a 70 kDa, glycosyl phosphatidylinositol (GPI)-anchored glycoprotein. CD73 is expressed on subsets of T and B lymphocytes, follicular dendritic cells, epithelial cells, endothelial cells and mesenchymal stem cells. Its expression on lymphocytes increases during T and B cell development. CD73 has enzymatic activity and catalyzes the dephosphorylation of adenosine monophosphate (AMP) converting it to adenosine. It has been suggested that CD73 can mediate costimulatory signals in T cell activation and adhesion of lymphocytes to endothelium.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD73
Applications Flow Cytometry
Classification Monoclonal
Clone AD2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description NT5E; 5' nucleotidase; 5'-NT; E5NT; Ecto-5'-nucleotidase; eN; eNT; NT; NT5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Pre-B leukemia cell line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.