Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD74, HLA-DQA1 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89022987 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-987 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-987 Supplier Abnova Corporation Supplier No. DI0228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

CD74 rabbit purified polyclonal antibody (20μg); HLA-DQA1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CD74 protein, and the other against the HLA-DQA1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen CD74
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene CD74
Gene Alias DHLAG/HLADG/Ia-GAMMA
Gene Symbols CD74
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 972/3117
Includes Antibody pair set content: 1. CD74 rabbit purified polyclonal antibody (20 ug) 2. HLA-DQA1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type CD74 & HLA-DQA1 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.