Learn More
Abcam Anti-CD80 Antibody, Rabbit Multiclonal, Human Mouse, 100 μL
Description
- Host species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Applications: Flow Cyt, IP, WB
- Species reactivity: Human, Mouse
- Purification: Affinity purification Protein A
- Storage buffer: pH 7.2 to 7.4, 0.01% Sodium azide, PBS, 40% Glycerol, 0.05% BSA
- Features: Knockout tested
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Human, Mouse |
| Buffer | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH7.2 to pH7.4 |
| Label or Dye | Unconjugated |
| Detection Location | In-Blot Detection, In-Cell Detection |
| Form | Liquid |
| Target Molecule | CD80 (Human, Mouse) |
| Cross Reactivity | Human, Mouse |
| Formulation | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH 7.2 to 7.4 |
| Quantity | 100 μL |
| For Use With (Application) | Western Blot, Flow Cytometry, IP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.