Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Mouse anti-Human, Unlabeled, Clone: JS-81, BD

Catalog No. BDB555675 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555675 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555675 Supplier BD Biosciences Supplier No. 555675
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

JS-81 identifies the newly clustered CD81 antigen, a 26kDa, type III transmembrane protein (also known as TAPA-1) that is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. MAbs to CD81 have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex which can include the Leu13 molecule, and the CD19 and CD21 molecules in B cells. MAbs to CD81 are reported to induce homotypic cell aggregation.

Host Species: Mouse
Clone: JS-81
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone JS-81
Concentration 0.5mg/mL
Conjugate Unconjugated
Description TAPA-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Human, Rabbit
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.